3-Amino-1H-1,2,4-triazole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR13640-250G OR13640-10G
APOSOR13640-250GEA 116 GBP
APOSOR13640-250G APOSOR13640-10G
3-Amino-1H-1,2,4-triazole
3-Amino-1H-1,2,4-triazole
Formula: C₂H₄N₄
Melting Pt: 149…153 °C
Density: 1.138 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00053362
CAS Number: 61-82-5
EINECS: 200-521-5
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR