Hydrobromic acid 48% in aqueous solution
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
OR13036-1KG
OR13036-5KG
APOSOR13036-1KGEA
27.3
GBP
APOSOR13036-1KG
APOSOR13036-5KG
Hydrobromic acid 48% in aqueous solution
Hydrogen bromide
Formula:
HBr Boiling Pt: 126…128 °C Melting Pt: –11 °C Density: 1.48 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011323 CAS Number: 10035-10-6 EINECS: 233-113-0 UN: 1788 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...