2-Naphthalenesulphonyl chloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR11219-100G OR11219-25G OR11219-500G
APOSOR11219-100GEA 22.9 GBP
APOSOR11219-100G APOSOR11219-25G APOSOR11219-500G
2-Naphthalenesulphonyl chloride
2-Naphthalenesulphonyl chloride
Formula: C₁₀H₇ClO₂S
Boiling Pt: 200…202 °C/13mmHg
Melting Pt: 74…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00004087
CAS Number: 93-11-8
EINECS: 202-219-9
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR