3-Aminobenzonitrile

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 3-Cyanoaniline, m-Aminobenzonitrile, m-Cyanoaniline

OR11110-100G OR11110-25G OR11110-5G
APOSOR11110-100GEA 103 GBP
APOSOR11110-100G APOSOR11110-25G APOSOR11110-5G
3-Aminobenzonitrile
3-Aminobenzonitrile
Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 289…290 °C
Melting Pt: 48…53 °C
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00007756
CAS Number: 2237-30-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR