3-Oxo-3,4-dihydro-2H-benzo[1,4]oxazine-6-sulphonyl chloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR103646-1G OR103646-250MG
APOSOR103646-1GEA 54.8 GBP
APOSOR103646-1G APOSOR103646-250MG
3-Oxo-3,4-dihydro-2H-benzo[1,4]oxazine-6-sulphonyl chloride
3-Oxo-3,4-dihydro-2H-benzo[1,4]oxazine-6-sulphonyl chloride
Formula: C₈H₆ClNO₄S
Melting Pt: 177.5…178.5 °C
Storage Temperature: Ambient
MDL Number: MFCD05664887
CAS Number: 31794-45-3
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR