Pyridazine-3-carboxylic acid 96%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
OR1005-10G
OR1005-1G
OR1005-5G
APOSOR1005-10GEA
221
GBP
APOSOR1005-10G
APOSOR1005-1G
APOSOR1005-5G
Pyridazine-3-carboxylic acid 96%
Pyridazine-3-carboxylic acid
Formula:
C₅H₄N₂O₂ MW: 124.10 g/mol Melting Pt: 190…195 °C Storage Temperature: Ambient |
MDL Number:
MFCD01646333 CAS Number: 2164-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...