Pyridine 99% ACS

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR10003-1L OR10003-4L OR10003-500ML
APOSOR10003-1LEA 145 GBP
APOSOR10003-1L APOSOR10003-4L APOSOR10003-500ML
Pyridine 99% ACS
Pyridine
Formula: C₅H₅N
Boiling Pt: 114…115 °C
Melting Pt: –42 °C
Density: 0.979 g/cm³ (25 °C)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR