4-(2-Chloroethyl)morpholine hydrochloride 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR0990-100G OR0990-25G
APOSOR0990-100GEA 21 GBP
APOSOR0990-100G APOSOR0990-25G
4-(2-Chloroethyl)morpholine hydrochloride 99%
4-(2-Chloroethyl)morpholine hydrochloride
Formula: C₆H₁₃Cl₂NO
Melting Pt: 180…186 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012797
CAS Number: 3647-69-6
EINECS: 222-881-2
UN: 2923
ADR: 8+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR