4-Piperidin-4-ylmorpholine 98+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Morpholinopiperidine

OR0953-1G OR0953-5G
APOSOR0953-1GEA 15.8 GBP
APOSOR0953-1G APOSOR0953-5G
4-Piperidin-4-ylmorpholine 98+%
4-Piperidin-4-ylmorpholine
Formula: C₉H₁₈N₂O
Boiling Pt: 100…115 °C/0.15mmHg
Melting Pt: 40…43 °C
Storage Temperature: Ambient
MDL Number: MFCD03274733
CAS Number: 53617-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR