2,6-Dihydroxybenzoic acid 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR0909-100G OR0909-25G OR0909-500G
APOSOR0909-100GEA 17.7 GBP
APOSOR0909-100G APOSOR0909-25G APOSOR0909-500G
2,6-Dihydroxybenzoic acid 98%
2,6-Dihydroxybenzoic acid
Formula: C₇H₆O₄
Melting Pt: 158 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002462
CAS Number: 303-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR