Rubidium dihydrogenarsenate 99,9%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

IN3024-25G IN3024-5G
APOSIN3024-25GEA 399 GBP
APOSIN3024-25G APOSIN3024-5G
Rubidium dihydrogenarsenate 99,9%
Rubidium dihydrogenarsenate
Formula: H₂AsO₄Rb
Melting Pt: 268 °C
Storage Temperature: Ambient
CAS Number: 13464-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR