Lead(II) sulphide 99.995%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

IN2293-100G IN2293-25G
APOSIN2293-100GEA 494 GBP
APOSIN2293-100G APOSIN2293-25G
Lead(II) sulphide 99.995%
Lead (II) sulphide
Formula: ̍PbS
Melting Pt: 1114 °C
Density: 7.5 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016280
CAS Number: 1314-87-0
EINECS: 215-246-6
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR