Lead molybdenum oxide 99,9%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IN2248-10G
IN2248-50G
APOSIN2248-10GEA
52.5
GBP
APOSIN2248-10G
APOSIN2248-50G
Lead molybdenum oxide 99,9%
Lead molybdenum oxide
Formula:
MoO₄Pb Melting Pt: 1060…1070 °C Density: 6.92 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016272 CAS Number: 10190-55-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...