Lead(II) fluoride 99.99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Lead difluoride

IN2239-250G IN2239-50G
APOSIN2239-250GEA 163 GBP
APOSIN2239-250G APOSIN2239-50G
Lead(II) fluoride 99.99%
Lead (II) fluoride
Formula: F₂Pb
MW: 245.2 g/mol
Boiling Pt: 1290 °C
Melting Pt: 855 °C
Density: 8.445 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011162
CAS Number: 7783-46-2
Merck Index: 12,05425

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR