Lead(II) chloride 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

IN2218-2KG IN2218-500G
APOSIN2218-2KGEA 214 GBP
APOSIN2218-2KG APOSIN2218-500G
Lead(II) chloride 99%
Lead (II) chloride
Formula: Cl₂Pb
Boiling Pt: 950 °C
Melting Pt: 501 °C
Density: 5.85 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011157
CAS Number: 7758-95-4
EINECS: 231-845-5
UN: 2291
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR