Arsenic(III) sulphide 99.999%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Arsenic trisulphide, Disrsenic trisulphide

IN1166-25G IN1166-5G
APOSIN1166-25GEA 805 GBP
APOSIN1166-25G APOSIN1166-5G
Arsenic(III) sulphide 99.999%
Arsenic (III) sulphide
Formula: As₂S₃
MW: 246.04 g/mol
Boiling Pt: 707 °C
Melting Pt: 300 °C
Density: 3.43 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003435
CAS Number: 1303-33-9
UN: 1557
ADR: 6.1 III
Merck Index: 12,00846

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR