PIPES dipotassium salt ≥95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BIP0706-100G BIP0706-500G
APOSBIP0706-100GEA 64.1 GBP
APOSBIP0706-100G APOSBIP0706-500G
PIPES dipotassium salt ≥95%
PIPES dipotassium salt

  • Buffer

Formula: C₈H₁₆N₂O₆S₂K₂
Storage Temperature: Ambient
CAS Number: 108321-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR