Pimaricin

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Natamycin

BIP0144-1G BIP0144-500MG BIP0144-5G
APOSBIP0144-1GEA 78.7 GBP
APOSBIP0144-1G APOSBIP0144-500MG APOSBIP0144-5G
Pimaricin
Pimaricin

Pimaricin is a polyene antifungal antibiotic produced by Streptomyces natalensis


  • Cell culture tested

Formula: C₃₃H₄₇NO₁₃
MW: 665.733 g/mol
Melting Pt: 280 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00135085
CAS Number: 7681-93-8
EINECS: 231-683-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR