Oligomycin A (from Streptomyces diastatochromogenes)
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
(1R,2'R,4E,5'S,6S,6'S,7R,8S,10R,11R,12S,14R,15S,16R,18E,20E,22R,25S,28S,29R)-22-Ethyl-7,11,14,15-tetrahydroxy-6'-((R)-2-hydroxypropyl)-5',6,8,10,12,14,16,28,29-nonamethyl-3',4',5',6'-tetrahydro-2,26-dioxaspiro[bicyclo[23.3.1]nonacosa[4,18,20]triene-27,2'-pyran]-3,9,13-trione
BIO1003-10MG
BIO1003-5MG
APOSBIO1003-10MGEA
279
GBP
APOSBIO1003-10MG
APOSBIO1003-5MG
Oligomycin A (from Streptomyces diastatochromogenes)
Oligomycin A
Practically free of homologs.
Formula:
C₄₅H₇₄O₁₁ MW: 791.07 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00065705 CAS Number: 579-13-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...