3'-Amino-2',3'-dideoxyguanosine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BIMN2003-100MG BIMN2003-1G BIMN2003-250MG
APOSBIMN2003-100MGEA 42 GBP
APOSBIMN2003-100MG APOSBIMN2003-1G APOSBIMN2003-250MG
3'-Amino-2',3'-dideoxyguanosine
3'-Amino-2',3'-dideoxyguanosine
Formula: C₁₀H₁₄N₆O₃
MW: 266.26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09750824
CAS Number: 66323-49-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR