Triclosan

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

BII1002-100G BII1002-25G
APOSBII1002-100GEA 94.9 GBP
APOSBII1002-100G APOSBII1002-25G
Triclosan
Triclosan

  • Antibodies

Formula: C₁₂H₇Cl₃O₂
Melting Pt: 56…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00800992
CAS Number: 3380-34-5
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR