(Indol-3-yl)acetic acid 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Heterauxin, Indol-3-ylacetic acid, IAA, 3-Indolylacetic acid

BII0601-100G BII0601-25G BII0601-500G
APOSBII0601-100GEA 52.5 GBP
APOSBII0601-100G APOSBII0601-25G APOSBII0601-500G
(Indol-3-yl)acetic acid 99%
(Indol-3-yl)acetic acid

  • Plant tissue culture tested

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 165…169 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005636
CAS Number: 87-51-4
EINECS: 201-748-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR