3,3'-Diaminobenzidine (DAB)

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: [1,1'-Biphenyl]-3,3',4,4'-tetraamine, DAB, 3,3',4,4'-Biphenyltetramin, Biphenyl-3,3',4,4'-tetrayltetraamine

BID2042-10G BID2042-5G
APOSBID2042-10GEA 30.5 GBP
APOSBID2042-10G APOSBID2042-5G
3,3'-Diaminobenzidine (DAB)
3,3'-Diaminobenzidine

Substrate for peroxidase. Also can be used for determination of selenium.

Formula: C₁₂H₁₄N₄
Melting Pt: 175…180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007725
CAS Number: 91-95-2
EINECS: 202-110-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR