3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine dihydrochloride xhydrate, TMB-HCl hydrate, Dihydrochloride TMB hydrate

BIB6275-10G BIB6275-5G
APOSBIB6275-10GEA 37.8 GBP
APOSBIB6275-10G APOSBIB6275-5G
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate
Formula: C₁₆H₂₀N₂·2HCl·xH₂O
MW: 313.27 g/mol (anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD00150104
CAS Number: 207738-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR