3,3',5,5'-Tetramethylbenzidine (TMB) free base

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine

BIB6274-100G BIB6274-10G BIB6274-50G
APOSBIB6274-100GEA 347 GBP
APOSBIB6274-100G APOSBIB6274-10G APOSBIB6274-50G
3,3',5,5'-Tetramethylbenzidine (TMB) free base
3,3',5,5'-Tetramethylbenzidine (TMB)
Formula: C₁₆H₂₀N₂
MW: 240.35 g/mol
Boiling Pt: 402
Melting Pt: 168…171 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007748
CAS Number: 54827-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR