Phytic acid, dodecasodium salt

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Inositol hexaphosphoric acid, dodecasodium salt, myo-Inositol hexakis(dihydrogen phosphate), dodecasodium salt

BIB6261-100G BIB6261-250G BIB6261-50G
APOSBIB6261-100GEA 69.3 GBP
APOSBIB6261-100G APOSBIB6261-250G APOSBIB6261-50G
Phytic acid, dodecasodium salt
Phytic acid, dodecasodium salt
Formula: C₆H₆Na₁₂O₂₄P₆
Storage Temperature: Ambient
MDL Number: MFCD00082310
CAS Number: 17211-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR