Imidazole 99+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine

BI7565-25G BI7565-5G
APOSBI7565-25GEA 48.3 GBP
APOSBI7565-25G APOSBI7565-5G
Imidazole 99+%
Imidazole

Biological buffer.


  • Assay (by titration) 99+%
  • Water (by Karl Fischer) <1.0%
  • White crystals

Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 235 °C
Melting Pt: 85…91 °C
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 3263
ADR: 8 II
Merck Index: 13,04935

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR