2-Fluoro-4-isopropoxyaniline 98%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC900145-1G
PC900145-5G
APOSPC900145-1GEA
163
GBP
APOSPC900145-1G
APOSPC900145-5G
2-Fluoro-4-isopropoxyaniline 98%
2-Fluoro-4-isopropoxyaniline
Formula:
C₉H₁₂FNO Storage Temperature: Ambient |
MDL Number:
MFCD15209543 CAS Number: 154080-04-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...