4-Fluoro-3-methoxybenzaldoxime

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC6396-1G PC6396-5G
APOSPC6396-1GEA 63 GBP
APOSPC6396-1G APOSPC6396-5G
4-Fluoro-3-methoxybenzaldoxime
4-Fluoro-3-methoxybenzaldoxime
Formula: C₈H₈FNO₂
MW: 169.153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412435
CAS Number: 886762-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR