1H-Perfluoroheptane 97%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7-Pentadecafluoroheptane, 1H-Pentadecafluoroheptane
PC6030-1G
PC6030-5G
PC6030-250MG
APOSPC6030-1GEA
55.7
GBP
APOSPC6030-1G
APOSPC6030-5G
APOSPC6030-250MG
1H-Perfluoroheptane 97%
1H-Perfluoroheptane
Formula:
C₇HF₁₅ Boiling Pt: 96…97 °C Density: 1.725 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00042108 CAS Number: 375-83-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...