[2-Bromo-6-fluoro-4-(trifluoromethyl)phenyl]methanol 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC57069-1G PC57069-250MG PC57069-5G
APOSPC57069-1GEA 372 GBP
APOSPC57069-1G APOSPC57069-250MG APOSPC57069-5G
[2-Bromo-6-fluoro-4-(trifluoromethyl)phenyl]methanol 95%
[2-Bromo-6-fluoro-4-(trifluoromethyl)phenyl]methanol
Formula: C₈H₅BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD31812094
CAS Number: 2091023-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR