1-Bromo-2-(bromomethyl)-3-fluoro-5-(trifluoromethyl)benzene 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC57068-1G PC57068-250MG
APOSPC57068-1GEA 411 GBP
APOSPC57068-1G APOSPC57068-250MG
1-Bromo-2-(bromomethyl)-3-fluoro-5-(trifluoromethyl)benzene 95%
1-Bromo-2-(bromomethyl)-3-fluoro-5-(trifluoromethyl)benzene
Formula: C₈H₄Br₂F₄
MW: 335.919 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31812116
CAS Number: 2091622-24-9
UN: 1760
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR