Trifluoromethyl trifluoromethanesulphonate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC57038-5G PC57038-25G
APOSPC57038-5GEA 343 GBP
APOSPC57038-5G APOSPC57038-25G
Trifluoromethyl trifluoromethanesulphonate
Trifluoromethyl trifluoromethanesulphonate
Formula: C₂F₆O₃S
Boiling Pt: 19…23 °C
Melting Pt: –108…–105 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04973017
CAS Number: 3582-05-6
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR