3-Chloro-5-fluoropicolinaldehyde 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC51357-1G PC51357-250MG
APOSPC51357-1GEA 1050 GBP
APOSPC51357-1G APOSPC51357-250MG
3-Chloro-5-fluoropicolinaldehyde 95%
3-Chloro-5-fluoropicolinaldehyde
Formula: C₆H₃ClFNO
Storage Temperature: Ambient
MDL Number: MFCD16606530
CAS Number: 1227563-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR