2-Fluoro-6-nitrophenylthiocyanate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC508071-1G PC508071-250MG
APOSPC508071-1GEA 760 GBP
APOSPC508071-1G APOSPC508071-250MG
2-Fluoro-6-nitrophenylthiocyanate
2-Fluoro-6-nitrophenylthiocyanate
Formula: C₇H₃FN₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD30723324
CAS Number: 21325-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR