2-Fluoro-4-(triFluoromethylthio)aniline 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC508067-100MG PC508067-250MG
APOSPC508067-100MGEA 449 GBP
APOSPC508067-100MG APOSPC508067-250MG
2-Fluoro-4-(triFluoromethylthio)aniline 95%
2-Fluoro-4-(triFluoromethylthio)aniline
Formula: C₇H₅F₄NS
Storage Temperature: Refrigerator
MDL Number: MFCD21807292
CAS Number: 596850-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR