3-Fluoro-4-nitrophenylthiocyanate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC508062-1G PC508062-250MG PC508062-5G
APOSPC508062-1GEA 760 GBP
APOSPC508062-1G APOSPC508062-250MG APOSPC508062-5G
3-Fluoro-4-nitrophenylthiocyanate
3-Fluoro-4-nitrophenylthiocyanate
Formula: C₇H₃FN₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD30723317
CAS Number: 2149598-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR