4-Fluoro-2-nitrophenylthiocyanate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC508035-1G PC508035-250MG PC508035-5G
APOSPC508035-1GEA 760 GBP
APOSPC508035-1G APOSPC508035-250MG APOSPC508035-5G
4-Fluoro-2-nitrophenylthiocyanate
4-Fluoro-2-nitrophenylthiocyanate
Formula: C₇H₃FN₂O₂S
MW: 198.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30723336
CAS Number: 21325-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR