2,4-Difluoro-3-(trifluoromethoxy)benzonitrile

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC508033-1G PC508033-250MG
APOSPC508033-1GEA 283 GBP
APOSPC508033-1G APOSPC508033-250MG
2,4-Difluoro-3-(trifluoromethoxy)benzonitrile
2,4-Difluoro-3-(trifluoromethoxy)benzonitrile
Formula: C₈H₂F₅NO
Storage Temperature: Ambient
MDL Number: MFCD28791003
CAS Number: 1806314-81-7
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR