2,4-Difluoro-3-(trifluoromethoxy)iodobenzene 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC508028-1G PC508028-250MG
APOSPC508028-1GEA 523 GBP
APOSPC508028-1G APOSPC508028-250MG
2,4-Difluoro-3-(trifluoromethoxy)iodobenzene 95%
2,4-Difluoro-3-(trifluoromethoxy)iodobenzene
Formula: C₇H₂F₅IO
Storage Temperature: Ambient
MDL Number: MFCD30723451
CAS Number: 2149602-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR