3-bromo-4-fluorophenylthiocyanate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC508025-1G PC508025-250MG PC508025-5G
APOSPC508025-1GEA 760 GBP
APOSPC508025-1G APOSPC508025-250MG APOSPC508025-5G
3-bromo-4-fluorophenylthiocyanate
3-bromo-4-fluorophenylthiocyanate
Formula: C₇H₃BrFNS
Storage Temperature: Ambient
MDL Number: MFCD30723328
CAS Number: 2149601-24-9
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR