3-Fluoro-2-(trifluoromethoxy)-6-nitroaniline

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC508018-1G PC508018-250MG
APOSPC508018-1GEA 957 GBP
APOSPC508018-1G APOSPC508018-250MG
3-Fluoro-2-(trifluoromethoxy)-6-nitroaniline
3-Fluoro-2-(trifluoromethoxy)-6-nitroaniline
Formula: C₇H₄F₄N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD30723447
CAS Number: 2149598-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR