2-Bromo-3’-(trifluoromethyl)propiophenone
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Bromo-1-[3-(trifluoromethyl)phenyl]propan-1-one
PC508014-1G
PC508014-250MG
PC508014-5G
APOSPC508014-1GEA
185
GBP
APOSPC508014-1G
APOSPC508014-250MG
APOSPC508014-5G
2-Bromo-3’-(trifluoromethyl)propiophenone
2-Bromo-3’-(trifluoromethyl)propiophenone
Formula:
C₁₀H₈BrF₃O MW: 281.069 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18392396 CAS Number: 38786-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...