2,4-Difluoro-3-(trifluoromethoxy)benzaldehyde 97%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
PC508013-1G
PC508013-250MG
APOSPC508013-1GEA
442
GBP
APOSPC508013-1G
APOSPC508013-250MG
2,4-Difluoro-3-(trifluoromethoxy)benzaldehyde 97%
2,4-Difluoro-3-(trifluoromethoxy)benzaldehyde
Formula:
C₈H₃F₅O₂ MW: 226.1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30723442 CAS Number: 2149602-63-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...