4-bromo-3-fluorophenylthiocyanate
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
PC508010-1G
PC508010-250MG
PC508010-5G
APOSPC508010-1GEA
760
GBP
APOSPC508010-1G
APOSPC508010-250MG
APOSPC508010-5G
4-bromo-3-fluorophenylthiocyanate
4-bromo-3-fluorophenylthiocyanate
Formula:
C₇H₃BrFNS Storage Temperature: Ambient |
MDL Number:
MFCD30723314 CAS Number: 2149601-22-7 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...