2,4-Difluoro-3-(trifluoromethoxy)nitrobenzene
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
PC508009-1G
PC508009-250MG
APOSPC508009-1GEA
286
GBP
APOSPC508009-1G
APOSPC508009-250MG
2,4-Difluoro-3-(trifluoromethoxy)nitrobenzene
2,4-Difluoro-3-(trifluoromethoxy)nitrobenzene
Formula:
C₇H₂F₅NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD28790867 CAS Number: 153338-22-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...