2-bromo-4-fluorophenylthiocyanate
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
PC508005-1G
PC508005-250MG
PC508005-5G
APOSPC508005-1GEA
760
GBP
APOSPC508005-1G
APOSPC508005-250MG
APOSPC508005-5G
2-bromo-4-fluorophenylthiocyanate
2-bromo-4-fluorophenylthiocyanate
Formula:
C₇H₃BrFNS Storage Temperature: Ambient |
MDL Number:
MFCD30723309 CAS Number: 2149597-67-9 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...