2,4-Difluoro-3-(trifluoromethoxy)aniline

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC508003-1G PC508003-250MG
APOSPC508003-1GEA 676 GBP
APOSPC508003-1G APOSPC508003-250MG
2,4-Difluoro-3-(trifluoromethoxy)aniline
2,4-Difluoro-3-(trifluoromethoxy)aniline
Formula: C₇H₄F₅NO
Storage Temperature: Ambient
MDL Number: MFCD19440811
CAS Number: 153338-24-0
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR