1H,1H-Perfluoropropane 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1,1,1,2,2,3-Hexafluoropropane (FC-236cb)

PC4786-25G PC4786-5G
APOSPC4786-25GEA 1040 GBP
APOSPC4786-25G APOSPC4786-5G
1H,1H-Perfluoropropane 97%
1H,1H-Perfluoropropane
Formula: C₃H₂F₆
Boiling Pt: –1.4 °C
Density: 1.32 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00153190
CAS Number: 677-56-5
UN: 3163
ADR: 2.2,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR