Sodium dichlorofluoroacetate 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC450243-1G PC450243-5G
APOSPC450243-1GEA 47.3 GBP
APOSPC450243-1G APOSPC450243-5G
Sodium dichlorofluoroacetate 98%
Sodium dichlorofluoroacetate
Formula: C₂Cl₂FO₂·Na
Storage Temperature: Ambient
MDL Number: MFCD02696369
CAS Number: 2837-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR